Learn More
Biomatik Corporation Human Endothelin Converting Enzyme 2 (ECE2) ELISA Kit
Supplier:  Biomatik Corporation EKU08428
Encompass_Preferred
Human Endothelin Converting Enzyme 2 (ECE2) ELISA Kit for detecting ECE2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.46-10000pg/ml. Sensitivity: 45.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: O60344. Gene ID: 110599583. Target Synonyms: Methyltransferase-like region. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7362