Learn More
Biomatik Corporation Human Enhancer Of Zeste Homolog 2 (EZH2) CLIA Kit
Supplier:  Biomatik Corporation EKU10030
Encompass_Preferred
Human Enhancer Of Zeste Homolog 2 (EZH2) CLIA Kit for detecting EZH2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q15910. Gene ID: 2146. Target Synonyms: ENX-1; KMT6; KMT6A; Histone-lysine N-methyltransferase EZH2; Lysine N-methyltransferase 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6396