Learn More
Biomatik Corporation Human Enhancer Of Zeste Homolog 1 (EZH1) ELISA Kit
Supplier:  Biomatik Corporation EKU03897
Encompass_Preferred
Human Enhancer Of Zeste Homolog 1 (EZH1) ELISA Kit for detecting EZH1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92800. Gene ID: 2145. Target Synonyms: KMT6B; ENX-2; Histone-lysine N-methyltransferase EZH1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6580