Learn More
Sino Biological Anti-Enolase 1/ENO1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11554RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ENO1 / Enolase 1 / alpha-enolase (rh ENO1 / Enolase 1 / alpha-enolase; Catalog#11554-H07E; P06733-1; Met1-Lys434). ENO1 / Enolase 1 / alpha-enolase specific IgG was purified by Human ENO1 / Enolase 1 / alpha-enolase affinity chromatography.
Catalog No. 50-240-8442