Learn More
Sino Biological Anti-Enolase 2/ENO2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13219MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NSE/ENO2 (rh NSE/ENO2; Catalog#13219-H08E; P09104; Met1-Leu434). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8989