Learn More
Sino Biological Anti-Enolase 2/ENO2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13219T12100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ENO2 (rh ENO2; Catalog#13219-H08E; P09104; Met1-Leu434). ENO2 specific IgG was purified by Human ENO2 affinity chromatography.
Catalog No. 50-240-8929