Learn More
Sino Biological Anti-ENO3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14270RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ENO3 / beta-enolase (rh ENO3; Catalog#14270-H07E; AAH17249.1; Met1-Lys434). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4140