Learn More
Biomatik Corporation Human Enolase 1 (ENO1) CLIA Kit
Supplier:  Biomatik Corporation EKU09896
Encompass_Preferred
Human Enolase 1 (ENO1) CLIA Kit for detecting ENO1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P06733. Gene ID: 2023. Target Synonyms: NNE; ENO1L1; MPB1; PPH; Alpha Enolase; Enolase 1; Phosphopyruvate hydratase; Plasminogen-binding protein; 2-phospho-D-glycerate hydro-lyase; C-myc promoter-binding. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8405