Learn More
Sino Biological Recombinant Human Urokinase/uPA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10815H08HA50
Encompass_Preferred
A DNA sequence encoding the human PLAU (NP_002649.1) (Met 1-Leu 431) with a C-terminal polyhistidine tag was expressed. The purified protein was activated by trypsin in vitro.
Catalog No. 50-224-7281