Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ENPP-1 (NP_006199) VersaClone cDNA

Catalog No. RDC2858
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2858 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2858 Supplier R&D Systems™ Supplier No. RDC2858
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Gene ENPP-1
Accession Number NP_006199
Gene ID (Entrez) 5167
Gene Alias ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, ENPP1, Ly-41 Antigen, M6S1, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPP1, NPPS, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1, PCA1ARHR2, PDNP1, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.