Learn More
Sino Biological Human ENPP2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11308MM13100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Autotaxin/ENPP2 (rh Autotaxin/ENPP2; Catalog#11308-H07H; AAH34961.1; Asp49-Ile863). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3602