Learn More
Sino Biological Human ENPP2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11308T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Autotaxin/ENPP2 (rh Autotaxin/ENPP2; Catalog#11308-H07H; AAH34961.1; Asp49-Ile863). Autotaxin/ENPP2 specific IgG was purified by Human Autotaxin/ENPP2 affinity chromatography.
Catalog No. 50-237-3694