Learn More
ACROBiosystems Human ENPP3 / CD203c Protein, His Tag
Catalog No. 502054950
Encompass_Preferred
Recombinant Protein;1MG;Human ENPP3, His Tag (EN3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 48 - Ile 875 (Accession # O14638-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 96.9 kDa.
Catalog No. 50-205-4950 Supplier ACROBiosystems Supplier No. EN3H52H41MG
Recombinant Protein;1MG;Human ENPP3, His Tag (EN3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 48 - Ile 875 (Accession # O14638-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 96.9 kDa.