Learn More
Sino Biological Recombinant Human CD203c/ENPP-3 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 19535H07H1
Encompass_Preferred
A DNA sequence encoding the Human ENPP3 (O14638) (Leu48-Ile875) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-241-2566