Learn More
Sino Biological Anti-ENPP-5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11519RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ENPP5 (rh ENPP5; Catalog#11519-H08H; Q9UJA9; Met1-Gly429). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4702