Learn More
Sino Biological Recombinant Human ENSA Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 14321H07EB100
Encompass_Preferred
A DNA sequence encoding the mature form of human ENSA (O43768-1) (Met1-Glu121) was expressed with a polyhistide tag at the N-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4138