Learn More
ATCC Human enterovirus 71; Strain: BrCr; Quantitative RNA
Supplier:  ATCC VR1775DQ
Encompass
Quantitative Genomic RNA from Human enterovirus 71 strain BrCr can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4394