Learn More
Sino Biological Recombinant Human CD39/ENTPD1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 16020H08B500
Encompass_Preferred
A DNA sequence encoding the human ENTPD1 (NP_001767.3) (Thr38-Val478) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4071