Learn More
Sino Biological Recombinant Human CD39L3/ENTPD3 Protein (His Tag)
Supplier:  Sino Biological 10909H08B50
Encompass_Preferred
A DNA sequence encoding the human ENTPD3 (O75355) extracellular domain (Gln 44-Pro 485) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-224-6691