Learn More
Sino Biological Anti-CD39L3/ENTPD3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10909RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ENTPD3 / NTPDase3 / CD39L3 (rh ENTPD3 / NTPDase3 / CD39L3; Catalog#10909-H08B; O75355; Gln44-Pro485). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8990