Learn More
Sino Biological Human ENTPD5 ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological 80387R07B150
Encompass_Preferred
Each vial contains 27 ng of recombinant Human ENTPD5. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
Catalog No. 50-237-4182