Learn More
Biomatik Corporation Human Envoplakin (EVPL) ELISA Kit
Supplier:  Biomatik Corporation EKU03909
Encompass_Preferred
Human Envoplakin (EVPL) ELISA Kit for detecting EVPL in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92817. Gene ID: 2125. Target Synonyms: EVPK; p210; IVL; 210 kDa cornified envelope precursor protein; 210 kDa paraneoplastic pemphigus antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6585