Learn More
Creative Biomart Alglucosidase alfa, Recombinant Human enzyme acid alpha-glucosidase (GAA)
Catalog No. 502084197
Encompass_Preferred
The product consists of the human enzyme acid alpha-glucosidase (GAA) which is essential for the degradation of glygogen to glucose in lysosomes. It is encoded by the most predominant of nine observed haplotypes of this gene. The product is produced by recombinant DNA technology in a Chinese hamster ovary cell line. The product degrades glycogen by catalyzing the hydrolysis of a-1,4- and a-1,6- glycosidic linkages of lysosomal glycogen. Structurally, the product is a glycoprotein with a calculated mass of 98,008 daltons for the 883 residue mature polypeptide chain, and a total mass of approximately 109,000 daltons, including carbohydrates.
Catalog No. 50-208-4197 Supplier Creative Biomart Supplier No. THP0146
The product consists of the human enzyme acid alpha-glucosidase (GAA) which is essential for the degradation of glygogen to glucose in lysosomes. It is encoded by the most predominant of nine observed haplotypes of this gene. The product is produced by recombinant DNA technology in a Chinese hamster ovary cell line. The product degrades glycogen by catalyzing the hydrolysis of a-1,4- and a-1,6- glycosidic linkages of lysosomal glycogen. Structurally, the product is a glycoprotein with a calculated mass of 98,008 daltons for the 883 residue mature polypeptide chain, and a total mass of approximately 109,000 daltons, including carbohydrates.