Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Eotaxin 3 ELISA Kit

Catalog No. EH171RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH171RBX5 5 x 96 Tests
EH171RBX10 10 x 96 Tests
EH171RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH171RBX5 Supplier Invitrogen™ Supplier No. EH171RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human Eotaxin 3 quantitates human Eotaxin 3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Eotaxin 3.

Eotaxin-3 is a small cytokine belonging to the CC chemokine family that is also called CCL26, Chemokine (C-C motif) ligand 26. Chemokines play pivotal roles in health and disease by controlling migration and tissue localization of specific types of cells expressing their cognate receptors. Eotaxin-3 is chemotactic for eosinophils and basophils and elicits its effects by binding to the cell surface chemokine receptor CCR3. It is expressed by several tissues including heart, lung and ovary, and in endothelial cells that have been stimulated with the cytokine interleukin 4. This gene for chemokine is located on human chromosome 7. Eotaxin-3 induced migration of eosinophils and basophils only at a 10-fold higher concentration than eotaxin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y258
Assay Range 28-7000 pg/mL
Assay Sensitivity 28 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CC chemokine IMAC; C-C motif chemokine 26; C-C motif chemokine ligand 26; Ccl26; Ccl26l; chemokine (C-C motif) ligand 26; chemokine N1; Eotaxin3; eotaxin-3; IMAC; Macrophage inflammatory protein 4-alpha; MGC126714; MIP-4a; MIP-4alpha; MIP-4-alpha; PRO242; SCYA26; SCYA26MIP-4-alpha; small inducible cytokine A26; small inducible cytokine subfamily A (Cys-Cys), member 26; small-inducible cytokine A26; Thymic stroma chemokine-1; TSC-1; UNQ216; UNQ216/PRO242CC chemokine IMAC; C-C motif chemokine 26; C-C motif chemokine ligand 26; Ccl26; Ccl26l; chemokine (C-C motif) ligand 26; chemokine N1; Eotaxin3; eotaxin-3; IMAC; Macrophage inflammatory protein 4-alpha; MGC126714; MIP-4a; MIP-4alpha; MIP-4-alpha; PRO242; SCYA26; SCYA26MIP-4-alpha; small inducible cytokine A26; small inducible cytokine subfamily A (Cys-Cys), member 26; small-inducible cytokine A26; Thymic stroma chemokine-1; TSC-1; UNQ216; UNQ216/PRO242;
Gene ID (Entrez) 10344
Gene Symbol CCL26
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Eotaxin 3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.