Learn More
BIOHIPPO Human EP2 Microsample
SDP
Catalog No. 5000315024
Encompass
Human EP2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EP2 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human EP2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human EP2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human EP2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15024 Supplier BIOHIPPO Supplier No. BHE15211379
Human EP2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EP2 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human EP2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human EP2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human EP2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color