Learn More
Sino Biological Recombinant Human EpCAM/TROP1 Protein (Site-Specific AF 488 Conjugation, ECD, His Tag)
Catalog No. 502993662
Encompass_Preferred
A DNA sequence encoding the Human EpCAM (NP_002345.1) (Met1-Lys265) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with AF 488 (Excitation Max. = 495 nm, Emission Max. = 519 nm).
Catalog No. 50-299-3662 Supplier Sino Biological Supplier No. 10694H86HSD100
A DNA sequence encoding the Human EpCAM (NP_002345.1) (Met1-Lys265) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with AF 488 (Excitation Max. = 495 nm, Emission Max. = 519 nm).