Learn More
Sino Biological Recombinant Human EpCAM/TROP1 Protein (Site-Specific PE Conjugation, ECD, His Tag)
Catalog No. 502992858
Encompass_Preferred
A DNA sequence encoding the Human EpCAM (NP_002345.1) (Met1-Lys265) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with PE (Excitation = 495-566 nm, Emission Max. = 575 nm).
Catalog No. 50-299-2858 Supplier Sino Biological Supplier No. 10694H86HSP25
A DNA sequence encoding the Human EpCAM (NP_002345.1) (Met1-Lys265) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with PE (Excitation = 495-566 nm, Emission Max. = 575 nm).