Learn More
Sino Biological Human EpCAM MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10694R028100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EpCAM / TROP1 (rh EpCAM; Catalog#10694-H08H; NP_002345.1; Met 1-Lys 265).
Catalog No. 50-237-2174