Learn More
Sino Biological Anti-EpCAM/TROP1 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10694MM06F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EpCAM / TROP1 / CD326 (rh EpCAM / TROP1 / CD326; Catalog#10694-H08H; NP_002345.1; Met 1-Lys 265) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8488