Learn More
Sino Biological Anti-EpCAM/TROP1 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10694MM06A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EpCAM / TROP-1 / TACSTD1 (rh EpCAM / TROP-1 / TACSTD1; Catalog#10694-H08H; NP_002345.1; Met1-Lys265) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0970