Learn More
Sino Biological Recombinant Anti-EpCAM/TROP1 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10694R028A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EpCAM / TROP1 / CD326 (rh EpCAM / TROP1 / CD326; Catalog#10694-H08H; NP_002345.1; Met 1-Lys 265) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0978