Learn More
Sino Biological Anti-EpCAM/TROP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10694T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human EpCAM / TROP-1 / TACSTD1 (rh EpCAM / TROP-1 / TACSTD1; Catalog#10694-H08H; NP_002345.1; Met1-Lys265). EpCAM / TROP-1 / TACSTD1 specific IgG was purified by Human EpCAM / TROP-1 / TACSTD1 affinity chromatography.
Catalog No. 50-241-1839