Learn More
Sino Biological Recombinant Human EpCAM/TROP1 Protein (ECD, hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 10694H41HB20
Encompass_Preferred
A DNA sequence encoding the human EPCAM (NP_002345.2) (Met1-Lys265) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5211