Learn More
ACROBiosystems ACROBiosystems Human EpCAM / TROP1 Protein, His Tag
Catalog No. 502106631
Encompass_Preferred
Recombinant Protein;100UG;Human EpCAM, His Tag (EPM-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6631 Supplier ACROBiosystems Supplier No. EPMH5223100UG
Recombinant Protein;100UG;Human EpCAM, His Tag (EPM-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.