Learn More
ACROBiosystems ACROBiosystems Human EpCAM / TROP1 Protein, Llama IgG2b Fc Tag
Catalog No. 502106921
Encompass_Preferred
Recombinant Protein;100UG;Human EpCAM, Llama IgG2b Fc Tag (EPM-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 55.2 kDa.
Catalog No. 50-210-6921 Supplier ACROBiosystems Supplier No. EPMH5259100UG
Recombinant Protein;100UG;Human EpCAM, Llama IgG2b Fc Tag (EPM-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 55.2 kDa.