Learn More
ACROBiosystems Human EpCAM / TROP1 Protein, Mouse IgG2a Fc Tag
Catalog No. 502055113
Encompass_Preferred
Recombinant Protein;100UG;Human EpCAM, Mouse IgG2a Fc Tag (EPM-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 54.3 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5113 Supplier ACROBiosystems Supplier No. EPMH5255100UG
Recombinant Protein;100UG;Human EpCAM, Mouse IgG2a Fc Tag (EPM-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 54.3 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.