Learn More
Sino Biological Recombinant Human EPCR Protein (hFc Tag), HPLC-verified 1
Supplier:  Sino Biological 13320H02H1
Encompass_Preferred
A DNA sequence encoding the human PROCR (AAH14451.1) (Met1-Thr209) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7299