Learn More
Abcam Human EphA2 ELISA Kit, 1 x 96 Tests
Catalog No. 502657495
Encompass_Preferred
Human EphA2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human EphA2 with a sensitivity of 0.88 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7495 Supplier Abcam Supplier No. AB2190511X96TESTS
Human EphA2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human EphA2 with a sensitivity of 0.88 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement