Learn More
BIORTUS USA INC Human EPHA2 Protein, 100UG
Catalog No. NC2953335
Encompass_Preferred
Gene Name: EPHA2. Uniprot: P29317. Construct: GST-TEV-GG-EPHA2(R561-I976 end). Expression Host: Insect cell. Purity: 90%.
Catalog No. NC2953335 Supplier BIORTUS USA INC Supplier No. BP1051202A
Gene Name: EPHA2. Uniprot: P29317. Construct: GST-TEV-GG-EPHA2(R561-I976 end). Expression Host: Insect cell. Purity: 90%.