Learn More
Sino Biological Recombinant Human EphA6 Protein (ECD, His Tag), HPLC-verified
Catalog No. 502993586
Encompass_Preferred
A DNA sequence encoding the Human EphA6 (Q9UF33) (Trp23-Val550) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3586 Supplier Sino Biological Supplier No. 16039H08H100
A DNA sequence encoding the Human EphA6 (Q9UF33) (Trp23-Val550) was expressed with a polyhistidine tag at the C-terminus.