Learn More
Sino Biological Anti-EphB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13925RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human EPHB3 / HEK2 (rh EPHB3 / HEK2; Catalog#13925-H20B1; P54753; Gln585-Val998). EPHB3 / HEK2 specific IgG was purified by Human EPHB3 / HEK2 affinity chromatography.
Catalog No. 50-240-9839