Learn More
Sino Biological Anti-EphB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13925RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human EPHB3 / HEK2 (aa 585-998) (rh EPHB3 / HEK2 (aa 585-998); Catalog#13925-H20B1; P54753; Gln585-Val998). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-7884