Learn More
Biotang Inc Human EphB4 / HTK Protein (His Tag),100ug, the extracellular domain (Met 1-Ala 539) of human EphB4 (NP_004435.3) precursor was expressed with a C-terminal polyhistidine tag in human cells.
Supplier:  Biotang Inc RPR9620
Encompass_Preferred
Human EphB4 / HTK Protein (His Tag),100ug, the extracellular domain (Met 1-Ala 539) of human EphB4 (NP_004435.3) precursor was expressed with a C-terminal polyhistidine tag in human cells.
Catalog No. 50-751-5482