Learn More
Sino Biological Recombinant Human EphB4 Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10235H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ala 539) of human EphB4 (NP_004435.3) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6981