Learn More
Sino Biological Recombinant Human EphB4 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10235H08HB100
Encompass_Preferred
A DNA sequence encoding the Human EPHB4 (NP_004435.3) (Met1-Ala539) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5417