Learn More
Sino Biological Recombinant Anti-EphB6 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10197R02620
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB6 / Eph Receptor B6 (rh EphB6 / Eph Receptor B6; Catalog#10197-H08H; NP_004436.1; Met1-Ser579).
Catalog No. 50-240-8804