Learn More
ACROBiosystems ACROBiosystems Human Ephrin-A1 / EFNA1 Protein, Fc Tag
Catalog No. 502106628
Encompass_Preferred
Recombinant Protein;1MG;Human Ephrin-A1, Fc Tag (EF1-H5251) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 182 (Accession # P20827-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.0 kDa. As a result of glycosylation, the protein migrates as 53-57 kDa under reducing (R) condition, and 100-115 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-210-6628 Supplier ACROBiosystems Supplier No. EF1H52511MG
Recombinant Protein;1MG;Human Ephrin-A1, Fc Tag (EF1-H5251) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 182 (Accession # P20827-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.0 kDa. As a result of glycosylation, the protein migrates as 53-57 kDa under reducing (R) condition, and 100-115 kDa under non-reducing (NR) condition (SDS-PAGE).