Learn More
Biomatik Corporation Human Ephrin A2 (EFNA2) ELISA Kit
Supplier:  Biomatik Corporation EKU08508
Encompass_Preferred
Human Ephrin A2 (EFNA2) ELISA Kit for detecting EFNA2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43921. Gene ID: 1943. Target Synonyms: ELF-1; EPLG6; HEK7-L; LERK6; EPH-related receptor tyrosine kinase ligand 6; HEK7 ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6910