Learn More
Biomatik Corporation Human Ephrin A5 (EFNA5) CLIA Kit
Supplier:  Biomatik Corporation EKU09973
Encompass_Preferred
Human Ephrin A5 (EFNA5) CLIA Kit for detecting EFNA5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P52803. Gene ID: 1946. Target Synonyms: AF1; EFL5; EPLG7; GLC1M; LERK7; RAGS; AL-1; EPH-related receptor tyrosine kinase ligand 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6497