Learn More
Biomatik Corporation Human Ephrin A5 (EFNA5) ELISA Kit
Supplier:  Biomatik Corporation EKU03923
Encompass_Preferred
Human Ephrin A5 (EFNA5) ELISA Kit for detecting EFNA5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52803. Gene ID: 1946. Target Synonyms: AF1; EFL5; EPLG7; GLC1M; LERK7; RAGS; AL-1; EPH-related receptor tyrosine kinase ligand 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8995